1-Bromo-3-isopropoxy-5-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

52897-500MG 52897-5G 52897-25G
77484-396EA 242.05 USD
77484-396 77494-900 77504-386
1-Bromo-3-isopropoxy-5-methylbenzene ≥95%
1-Bromo-3-isopropoxy-5-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19373758
CAS Number: 1268150-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR