(3-(Difluoromethoxy)-5-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

52895-500MG 52895-5G
77484-392EA 546.93 USD
77484-392 77494-896
(3-(Difluoromethoxy)-5-methylphenyl)boronic acid ≥97%
(3-(Difluoromethoxy)-5-methylphenyl)boronic acid

Precaución: For research use only

Formula: C₈H₉BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34756954
CAS Number: 2765315-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR