4-Chloro-2-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52827-500MG 52827-5G
77484-388EA 528.39 USD
77484-388 77494-890
4-Chloro-2-methoxy-5-methylbenzoic acid ≥97%
4-Chloro-2-methoxy-5-methylbenzoic acid

Precaución: For research use only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999871
CAS Number: 1368280-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR