5-(Hydroxymethyl)-2-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

52727-500MG 52727-5G
77484-380EA 555.17 USD
77484-380 77494-882
5-(Hydroxymethyl)-2-methylbenzonitrile ≥97%
5-(Hydroxymethyl)-2-methylbenzonitrile

Precaución: For research use only

Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394342
CAS Number: 315188-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR