2-Ethoxy-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

52603-500MG 52603-5G
77484-372EA 342.99 USD
77484-372 77494-874
2-Ethoxy-5-(trifluoromethoxy)benzoic acid ≥97%
2-Ethoxy-5-(trifluoromethoxy)benzoic acid

Precaución: For research use only

Formula: C₁₀H₉F₃O₄
MW: 250.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06208286
CAS Number: 773109-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR