Methyl(2,3,4-trifluorophenyl)sulfane ≥97%

Supplier: AOBChem USA

52480-5G 52480-25G
77494-868EA 249.26 USD
77494-868 77504-368
Methyl(2,3,4-trifluorophenyl)sulfane ≥97%
Methyl(2,3,4-trifluorophenyl)sulfane

Precaución: For research use only

Formula: C₇H₅F₃S
MDL Number: MFCD33404071
CAS Number: 2635937-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR