3-isopropyl-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52404-500MG 52404-5G 52404-25G
77484-366EA 299.73 USD
77484-366 77494-862 77504-364
3-isopropyl-5-methylbenzaldehyde ≥97%
3-isopropyl-5-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD12547789
CAS Number: 1208075-26-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR