(4-Bromo-1-fluoronaphthalen-2-yl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

52340-500MG 52340-5G 52340-25G
77484-362EA 88.58 USD
77484-362 77494-858 77504-358
(4-Bromo-1-fluoronaphthalen-2-yl)(methyl)sulfane ≥97%
(4-Bromo-1-fluoronaphthalen-2-yl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₁H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD33404046
CAS Number: 2624417-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR