3,3-Difluoro-5-methoxyindolin-2-one ≥95%

Supplier: AOBChem USA

Warning

52310-500MG 52310-5G 52310-25G
77484-358EA 284.28 USD
77484-358 77494-854 77504-356
3,3-Difluoro-5-methoxyindolin-2-one ≥95%
3,3-Difluoro-5-methoxyindolin-2-one

Precaución: For research use only

Formula: C₉H₇F₂NO₂
MW: 199.154 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22547890
CAS Number: 1360943-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR