(2-Chloro-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

52270-500MG 52270-5G 52270-25G
77484-354EA 296.64 USD
77484-354 77494-850 77504-354
(2-Chloro-5-(trifluoromethoxy)phenyl)(methyl)sulfane ≥95%
(2-Chloro-5-(trifluoromethoxy)phenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₆ClF₃OS
MW: 242.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756893
CAS Number: 2387119-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR