2-Bromo-2,2-difluoro-1-thiomorpholinoethanone ≥97%

Supplier: AOBChem USA

Warning

52253-500MG 52253-5G 52253-25G
77484-350EA 263.68 USD
77484-350 77494-844 77504-348
2-Bromo-2,2-difluoro-1-thiomorpholinoethanone ≥97%
2-Bromo-2,2-difluoro-1-thiomorpholinoethanone

Precaución: For research use only

Formula: C₆H₈BrF₂NOS
MW: 260.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD33404031
CAS Number: 1865110-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR