3,3-Difluoro-5-methylindolin-2-one ≥97%

Supplier: AOBChem USA

Warning

52238-500MG 52238-5G 52238-25G
77484-346EA 244.11 USD
77484-346 77494-840 77504-344
3,3-Difluoro-5-methylindolin-2-one ≥97%
3,3-Difluoro-5-methylindolin-2-one

Precaución: For research use only

Formula: C₉H₇F₂NO
MW: 183.155 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22556867
CAS Number: 221665-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR