Methyl 2-chloro-3-methoxyisonicotinate ≥97%

Supplier: AOBChem USA

Warning

52094-500MG 52094-5G 52094-25G
77484-342EA 300.76 USD
77484-342 77494-836 77504-340
Methyl 2-chloro-3-methoxyisonicotinate ≥97%
Methyl 2-chloro-3-methoxyisonicotinate

Precaución: For research use only

Formula: C₈H₈ClNO₃
MW: 201.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698149
CAS Number: 1214387-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR