3,4-Difluoro-2-methoxy-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51955-500MG 51955-5G
77484-338EA 408.91 USD
77484-338 77494-832
3,4-Difluoro-2-methoxy-5-(methylthio)benzaldehyde ≥97%
3,4-Difluoro-2-methoxy-5-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₈F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD34756860
CAS Number: 2807449-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR