2-Benzyl-4-bromo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

51790-500MG 51790-5G
77484-334EA 590.19 USD
77484-334 77494-828
2-Benzyl-4-bromo-1-methoxybenzene ≥97%
2-Benzyl-4-bromo-1-methoxybenzene

Precaución: For research use only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902975
CAS Number: 41876-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR