2-Chloro-3-fluoro-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

51738-500MG 51738-5G
77484-330EA 538.69 USD
77484-330 77494-824
2-Chloro-3-fluoro-4-methylbenzaldehyde ≥97%
2-Chloro-3-fluoro-4-methylbenzaldehyde

Precaución: For research use only

Formula: C₈H₆ClFO
MW: 172.584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751749
CAS Number: 1785126-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR