1,3-Dibromo-5-isopropyl-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51642-500MG 51642-5G 51642-25G
77484-326EA 108.15 USD
77484-326 77494-820 77504-328
1,3-Dibromo-5-isopropyl-2-methylbenzene ≥97%
1,3-Dibromo-5-isopropyl-2-methylbenzene

Precaución: For research use only

Formula: C₁₀H₁₂Br₂
MW: 292.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403986
CAS Number: 2624417-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR