1-Bromo-3,4-dichloro-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

51566-500MG 51566-5G 51566-25G
77484-322EA 293.55 USD
77484-322 77494-816 77504-326
1-Bromo-3,4-dichloro-2-methylbenzene ≥95%
1-Bromo-3,4-dichloro-2-methylbenzene

Precaución: For research use only

Formula: C₇H₅BrCl₂
Storage Temperature: Ambient
MDL Number: MFCD28737862
CAS Number: 1805023-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR