3-Isopropyl-5-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

51463-500MG 51463-5G 51463-25G
77484-314EA 253.38 USD
77484-314 77494-808 77504-322
3-Isopropyl-5-(methylthio)benzoic acid ≥97%
3-Isopropyl-5-(methylthio)benzoic acid

Precaución: For research use only

Formula: C₁₁H₁₄O₂S
MW: 210.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902941
CAS Number: 2586126-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR