Methyl 2,4-difluoro-3-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

51418-500MG 51418-5G 51418-25G
77484-310EA 120.51 USD
77484-310 77494-804 77504-318
Methyl 2,4-difluoro-3-methoxybenzoate ≥95%
Methyl 2,4-difluoro-3-methoxybenzoate

Precaución: For research use only

Formula: C₉H₈F₂O₃
MW: 202.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520396
CAS Number: 194804-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR