2-Fluoro-1-isopropoxy-4-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

51367-500MG 51367-5G
77484-302EA 342.99 USD
77484-302 77494-796
2-Fluoro-1-isopropoxy-4-(trifluoromethyl)benzene ≥97%
2-Fluoro-1-isopropoxy-4-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₁₀H₁₀F₄O
MW: 222.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23113825
CAS Number: 1394932-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR