1-Bromo-3,5-dichloro-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

51318-500MG 51318-5G 51318-25G
77484-298EA 203.94 USD
77484-298 77494-792 77504-310
1-Bromo-3,5-dichloro-2-ethoxybenzene ≥97%
1-Bromo-3,5-dichloro-2-ethoxybenzene

Precaución: For research use only

Formula: C₈H₇BrCl₂O
MW: 269.951 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29044108
CAS Number: 1010390-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR