2-(Benzyloxy)-1-bromo-3,5-dichlorobenzene ≥97%

Supplier: AOBChem USA

Warning

51313-500MG 51313-5G 51313-25G
77484-294EA 141.11 USD
77484-294 77494-788 77504-306
2-(Benzyloxy)-1-bromo-3,5-dichlorobenzene ≥97%
2-(Benzyloxy)-1-bromo-3,5-dichlorobenzene

Precaución: For research use only

Formula: C₁₃H₉BrCl₂O
MW: 332.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33403968
CAS Number: 2624417-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR