1-Ethoxy-3-fluoro-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

51309-5G 51309-25G
77494-784EA 255.44 USD
77494-784 77504-304
1-Ethoxy-3-fluoro-5-(trifluoromethyl)benzene ≥97%
1-Ethoxy-3-fluoro-5-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₉H₈F₄O
MDL Number: MFCD34530087
CAS Number: 2386302-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR