1-Chloro-2,4-difluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

51257-500MG 51257-5G 51257-25G
77484-288EA 141.11 USD
77484-288 77494-780 77504-300
1-Chloro-2,4-difluoro-3-methylbenzene ≥97%
1-Chloro-2,4-difluoro-3-methylbenzene

Precaución: For research use only

Formula: C₇H₅ClF₂
MW: 162.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922596
CAS Number: 1208075-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR