2-(4-Bromo-2,6-dimethoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

51182-500MG 51182-5G 51182-25G
77484-284EA 60.77 USD
77484-284 77494-776 77504-298
2-(4-Bromo-2,6-dimethoxyphenyl)-1,3-dioxolane ≥95%
2-(4-Bromo-2,6-dimethoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₃BrO₄
Storage Temperature: Ambient
MDL Number: MFCD33403958
CAS Number: 2624416-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR