1-(2,3-Difluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

51154-500MG 51154-5G
77484-276EA 322.39 USD
77484-276 77494-768
1-(2,3-Difluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥97%
1-(2,3-Difluoropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₇H₂F₅NO
MW: 211.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31567767
CAS Number: 2353676-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR