1-(3-Bromo-5-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

51138-500MG 51138-5G
77484-272EA 497.49 USD
77484-272 77494-764
1-(3-Bromo-5-fluoropyridin-4-yl)-2,2,2-trifluoroethanone ≥95%
1-(3-Bromo-5-fluoropyridin-4-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₇H₂BrF₄NO
Storage Temperature: Ambient
MDL Number: MFCD34813709
CAS Number: 2144049-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR