2,2,2-Trifluoro-1-(3-fluoropyridin-4-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

51127-500MG 51127-5G
77484-268EA 578.86 USD
77484-268 77494-760
2,2,2-Trifluoro-1-(3-fluoropyridin-4-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(3-fluoropyridin-4-yl)ethanone

Precaución: For research use only

Formula: C₇H₃F₄NO
MW: 193.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189068
CAS Number: 1060802-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR