1-(5-Bromo-2-fluoropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

50946-500MG 50946-5G
77484-264EA 621.09 USD
77484-264 77494-756
1-(5-Bromo-2-fluoropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%
1-(5-Bromo-2-fluoropyridin-3-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₇H₂BrF₄NO
MW: 271.994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28131837
CAS Number: 1824063-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR