1-(2,6-Difluoropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

50917-500MG 50917-5G 50917-25G
77484-260EA 161.71 USD
77484-260 77494-752 77504-290
1-(2,6-Difluoropyridin-3-yl)-2,2,2-trifluoroethanone ≥95%
1-(2,6-Difluoropyridin-3-yl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₇H₂F₅NO
MW: 211.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26958716
CAS Number: 1707361-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR