2,2,2-Trifluoro-1-(5-fluoropyridin-3-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

50878-500MG 50878-5G
77484-256EA 637.57 USD
77484-256 77494-748
2,2,2-Trifluoro-1-(5-fluoropyridin-3-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(5-fluoropyridin-3-yl)ethanone

Precaución: For research use only

Formula: C₇H₃F₄NO
MW: 193.098 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189067
CAS Number: 1060802-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR