2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

50863-500MG 50863-5G 50863-25G
77484-248EA 118.45 USD
77484-248 77494-740 77504-282
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(pyridin-3-yl)ethanone

Precaución: For research use only

Formula: C₇H₄F₃NO
MW: 175.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799596
CAS Number: 33284-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR