5-Bromo-2-fluoro-1-iodo-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

50796-500MG 50796-5G 50796-25G
77484-244EA 162.74 USD
77484-244 77494-736 77504-280
5-Bromo-2-fluoro-1-iodo-3-methylbenzene ≥95%
5-Bromo-2-fluoro-1-iodo-3-methylbenzene

Precaución: For research use only

Formula: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD28739625
CAS Number: 1627894-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR