(4-Chloro-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50755-500MG 50755-5G 50755-25G
77484-240EA 161.71 USD
77484-240 77494-732 77504-276
(4-Chloro-2-fluoro-5-methylphenyl)(methyl)sulfane ≥97%
(4-Chloro-2-fluoro-5-methylphenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752238
CAS Number: 1805954-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR