2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

50742-500MG 50742-5G 50742-25G
77484-236EA 230.72 USD
77484-236 77494-728 77504-272
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-6-hydroxy-4-(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₈H₄F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD27975474
CAS Number: 1823365-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR