(5-(Difluoromethoxy)-2-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

50609-500MG 50609-5G
77484-224EA 409.94 USD
77484-224 77494-716
(5-(Difluoromethoxy)-2-methylphenyl)boronic acid ≥97%
(5-(Difluoromethoxy)-2-methylphenyl)boronic acid

Precaución: For research use only

Formula: C₈H₉BF₂O₃
MW: 201.963 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756705
CAS Number: 2765316-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR