2,2,2-Trifluoro-1-(5-methylpyridin-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

50352-500MG 50352-5G 50352-25G
77484-212EA 111.24 USD
77484-212 77494-704 77504-262
2,2,2-Trifluoro-1-(5-methylpyridin-2-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(5-methylpyridin-2-yl)ethanone

Precaución: For research use only

Formula: C₈H₆F₃NO
MW: 189.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188989
CAS Number: 1060801-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR