(3-Bromo-2-ethoxy-6-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

50174-500MG 50174-5G 50174-25G
77484-208EA 95.79 USD
77484-208 77494-700 77504-260
(3-Bromo-2-ethoxy-6-fluorophenyl)(methyl)sulfane ≥97%
(3-Bromo-2-ethoxy-6-fluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34756694
CAS Number: 2706995-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR