4-(4-Bromo-2-(trifluoromethyl)phenoxy)benzonitrile ≥95%

Supplier: AOBChem USA

50008-5G 50008-25G
77494-692EA 234.84 USD
77494-692 77504-254
4-(4-Bromo-2-(trifluoromethyl)phenoxy)benzonitrile ≥95%
4-(4-Bromo-2-(trifluoromethyl)phenoxy)benzonitrile

Precaución: For research use only

Formula: C₁₄H₇BrF₃NO
MDL Number: MFCD34756669
CAS Number: 2755722-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR