Ethyl difluoro(pyridin-2-yl)acetate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2,2-difluoro-2-(pyridin-2-yl)acetate

49904-5G 49904-25G
77494-686EA 233.81 USD
77494-686 77504-250
Ethyl difluoro(pyridin-2-yl)acetate ≥95%
Ethyl difluoro(pyridin-2-yl)acetate

Precaución: For research use only

Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06858718
CAS Number: 267876-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR