2-(3,5-Dibromo-4-ethoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

49761-500MG 49761-5G 49761-25G
77484-196EA 296.64 USD
77484-196 77494-682 77504-246
2-(3,5-Dibromo-4-ethoxyphenyl)-1,3-dioxolane ≥97%
2-(3,5-Dibromo-4-ethoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₁H₁₂Br₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34756630
CAS Number: 2901106-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR