1-Bromo-4-chloro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49701-500MG 49701-5G 49701-25G
77484-192EA 152.44 USD
77484-192 77494-678 77504-244
1-Bromo-4-chloro-2-isopropoxybenzene ≥97%
1-Bromo-4-chloro-2-isopropoxybenzene

Precaución: For research use only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28356429
CAS Number: 1337606-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR