1,4-Dibromo-2-fluoro-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

49601-500MG 49601-5G
77484-188EA 367.71 USD
77484-188 77494-674
1,4-Dibromo-2-fluoro-5-(trifluoromethyl)benzene ≥95%
1,4-Dibromo-2-fluoro-5-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₇H₂Br₂F₄
MW: 321.892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28784686
CAS Number: 1806353-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR