1-Bromo-2-fluoro-3,5-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49464-500MG 49464-5G 49464-25G
77484-184EA 244.11 USD
77484-184 77494-670 77504-240
1-Bromo-2-fluoro-3,5-dimethoxybenzene ≥97%
1-Bromo-2-fluoro-3,5-dimethoxybenzene

Precaución: For research use only

Formula: C₈H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738435
CAS Number: 1804403-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR