(3-Bromo-2-chloro-5-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

49384-500MG 49384-5G 49384-25G
77484-176EA 237.93 USD
77484-176 77494-662 77504-232
(3-Bromo-2-chloro-5-fluorophenyl)boronic acid ≥97%
(3-Bromo-2-chloro-5-fluorophenyl)boronic acid

Precaución: For research use only

Formula: C₆H₄BBrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD32660673
CAS Number: 2379538-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR