2-(3-Bromo-2-chloro-5-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

49379-500MG 49379-5G 49379-25G
77484-172EA 182.31 USD
77484-172 77494-658 77504-228
2-(3-Bromo-2-chloro-5-fluorophenyl)-1,3-dioxolane ≥97%
2-(3-Bromo-2-chloro-5-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34756583
CAS Number: 2807455-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR