5-Bromo-2-(4-chlorophenoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

49192-500MG 49192-5G 49192-25G
77484-168EA 182.31 USD
77484-168 77494-652 77504-224
5-Bromo-2-(4-chlorophenoxy)pyridine ≥97%
5-Bromo-2-(4-chlorophenoxy)pyridine

Precaución: For research use only

Formula: C₁₁H₇BrClNO
MW: 284.536 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14652052
CAS Number: 28231-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR