1,3,5-trifluoro-2-isopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

49153-500MG 49153-5G
77484-164EA 434.66 USD
77484-164 77494-648
1,3,5-trifluoro-2-isopropylbenzene ≥95%
1,3,5-trifluoro-2-isopropylbenzene

Precaución: For research use only

Formula: C₉H₉F₃
MW: 174.163 g/mol
Storage Temperature: Ambient
CAS Number: 1535318-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR