(4-Ethoxy-2,3-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

49148-500MG 49148-5G
77484-160EA 374.92 USD
77484-160 77494-644
(4-Ethoxy-2,3-dimethylphenyl)methanol ≥97%
(4-Ethoxy-2,3-dimethylphenyl)methanol

Precaución: For research use only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902798
CAS Number: 1504246-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR